Please login to your online account to display your discounted pricing

American Educational Products Density Kit


Learn measurement, density, and the metric system

Manufacturer: american educational products  3131

 View more versions of this product

Catalog No. S53006

  • / Each
 

Description & Specifications

Specifications

Grade 6 to 12
Includes 3 x 1 x 0.25 in. Aluminum bar, two 0.5 in. Aluminum cubes, Steel sphere, Glass sphere, 15cm scale
Product Type Density kit
For Use With (Application) To learn measurement, the metric system, how to use the formula: Density=Mass/Volume