missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Denhardt's solution (100X)

Specifications

Chemical Name or Material Denhardt's solution
MDL Number MFCD00717549
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62416AD View Documents Alfa Aesar™
J62416AD
50mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ62416AE View Documents Alfa Aesar™
J62416AE
100mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Denhardt's solution
MFCD00717549