missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dendritic Cell Marker (33D1) Rat anti-Mouse, PerCP-eFluor 710, Clone: 33D1, eBioscience™

Rat Monoclonal Antibody

$187.05 - $433.55

Specifications

Antigen Dendritic Cell Marker (33D1)
Clone 33D1
Host Species Rat
Gene Alias DC Marker, DC inhibitor receptor 2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123854 Life Technologies
LS46588480
25μg Each for $187.05
501123855 Life Technologies
LS46588482
100μg Each for $433.55
Specifications

Description

The 33D1 monoclonal antibody reacts with a mouse dendritic cell (DC)-specific surface marker, DCIR2. The nature and biological activity of DCIR2 is unknown. DCIR2 has been reported on a variety of dendritic cell subpopulations from mouse thymus, spleen, lymph node, and Peyer’s patch. Bone marrow dendritic cells require GM-CSF to express DCIR2, and this expression is downregulated in the presence of IL-4. DCIR2 has been detected in vivo in brain dendritic cells post infection with Toxoplasma gondii.
Description & Specifications

Specifications

Dendritic Cell Marker (33D1)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
33D1
DC Marker, DC inhibitor receptor 2
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL