Please login to your online account to display your discounted pricing

Dendritic Cell Marker (33D1) Rat anti-Mouse, eFluor 660, Clone: 33D1, eBioscience™

Rat Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen Dendritic Cell Marker (33D1)
Clone 33D1
Host Species Rat
Gene Alias DC Marker, DC inhibitor receptor 2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124405

 
life technologies
LS50588480
25μg Each for $143.55

501124406

 
life technologies
LS50588482
100μg Each for $317.55
Description & Specifications

Specifications

Antigen Dendritic Cell Marker (33D1)
Clone 33D1
Host Species Rat
Gene Alias DC Marker, DC inhibitor receptor 2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 33D1 monoclonal antibody reacts with a mouse dendritic cell (DC)-specific surface marker, DCIR2. The nature and biological activity of DCIR2 is unknown. DCIR2 has been reported on a variety of dendritic cell subpopulations from mouse thymus, spleen, lymph node, and Peyer’s patch. Bone marrow dendritic cells require GM-CSF to express DCIR2, and this expression is downregulated in the presence of IL-4. DCIR2 has been detected in vivo in brain dendritic cells post infection with Toxoplasma gondii.