missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Denatured calf thymus DNA, phenol extracted

Chemical Identifiers

Chemical Name or Material Denatured calf thymus DNA, phenol extracted
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62977FPL SDS Alfa Aesar™
J62977FPL
20mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ62977MA SDS Alfa Aesar™
J62977MA
10mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Denatured calf thymus DNA, phenol extracted is been used in different nucleic acid related biochemical, molecular biological, cell biological, and immunological research studies. Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard for DNA quantification by fluorescent assay. It was used as reference to determine the DNA content in cartilage tissue digest samples.

Solubility
It is soluble in water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

Denatured calf thymus DNA, phenol extracted

Specifications

20mg
Liquid
Denatured calf thymus DNA, phenol extracted