DemoSlide
No need to bother with wet-mount slide preparation.
Manufacturer:  Fisher Scientific GSMSA
 View more versions of this product
Catalog No. S16441
Description
Description
  • Simply pipet or pour 5mL of live protozoa into a DemoSlide tube and view under your microscope
  • No spills, broken glass slides or evaporation — just easy-to-use tubes
  • Uses the Davstar Cen-Slide™
Two options:
  • Package of 120 DemoSlide tubes and caps — with this option, we recommend purchasing at least several sets of DemoSlide microscope stage adapters
  • Complete DemoSlide Kit: Includes 120 tubes and caps, stage adapter and a 12-place tube rack
Specifications
Specifications
Plastic
2/Pk.
Documents