Deluxe Inclined Plane Accessories
Manufacturer:  Fisher Science Education S40915
 View more versions of this product
Catalog No. S40915
Description
Description
Friction Block
  • Made of smooth finished maple or birch
  • With hook
  • Dimensions: 6.25 × 3 × 0.75 in.(15.9 × 7.6 × 1.9cm)
Friction Box
  • Open-top Baltic birch wooden box
  • Use to determine coefficient of static and kinetic friction
  • Features a hook at one end and two rubber strips secured to the bottom
  • Dimensions: 6.125 × 3.125 × 2.125 in. (15.6 × 7.9 × 5.4cm)
Specifications
Specifications
Friction Box
Ply Hardwood
Deluxe inclined plane
Three friction surfaces: two of smooth sanded Wood, one with Leather strips attached by adhesive