Please login to your online account to display your discounted pricing

Alfa Aesar™ Deltorphin I

For cellular and molecular biology applications

Specifications

Product Type Deltorphin I
Molecular Weight 768.87
Storage Requirements -30° to -10°C
Sequence Tyr-D-Ala-Phe-Asp-Val-Val-Gly-NH2
For Use With (Application) Selective delta-opioid receptor agonist
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60952LB0

 
alfa aesar™
J60952LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ60952MCR

 
alfa aesar™
J60952MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Deltorphin I
Molecular Weight 768.87
Storage Requirements -30° to -10°C
Sequence Tyr-D-Ala-Phe-Asp-Val-Val-Gly-NH2
For Use With (Application) Selective delta-opioid receptor agonist
Format Powder
Common Name Deltorphin C
Regulatory Status RUO
Purification Method HPLC