missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ delta-Valerolactam, 98+%

Manufacturer:  Alfa Aesar™ A1227114

 View more versions of this product

Catalog No. AAA1227114



Description

Description

Applications
delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

delta-Valerolactam
255°C to 257°C
25g
C5H9NO
106434
XUWHAWMETYGRKB-UHFFFAOYSA-N
piperidin-2-one
12665
99.13
147°C (296°F)
35°C to 39°C
675-20-7
MFCD00006037
2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2
C1CCNC(=O)C1
99.133
CHEBI:77761
≥98%

Chemical Identifiers

675-20-7
99.133
XUWHAWMETYGRKB-UHFFFAOYSA-N
12665
piperidin-2-one
C5H9NO
MFCD00006037
2-piperidone, 2-piperidinone, delta-valerolactam, valerolactim, 5-pentanolactam, piperidinone, piperidon, piperidone, alpha-piperidone, piperidone-2
CHEBI:77761
C1CCNC(=O)C1
Safety and Handling

Safety and Handling

RTECSNumber : TO0110000

TSCA : TSCA

SDS Documents