Please login to your online account to display your discounted pricing

Alfa Aesar™ delta-Tetradecanolactone, 98%

$59.07 - $628.32

Chemical Identifiers

Chemical Name or Material delta-Tetradecanolactone
CAS 2721-22-4
Molecular Formula C14H28O2
Formula Weight 226.36
MDL Number MFCD00059809
Synonym 5-Hydroxytetradecanoic acid lactone; delta-Tetradecalactone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2479414

  SDS
alfa aesar™
B2479414
25g Each for $59.07

AAB2479422

  SDS
alfa aesar™
B2479422
100g Each for $171.77

AAB2479436

  SDS
alfa aesar™
B2479436
500g Each for $628.32
Description & Specifications

Specifications

Formula Weight 226.36
Quantity 25g
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 5-Hydroxytetradecanoic acid lactone; delta-Tetradecalactone
Flash Point >110°C (230°F)
Molecular Formula C14H28O2
Melting Point 130° to 135°C
MDL Number MFCD00059809
Chemical Name or Material delta-Tetradecanolactone
TSCA TSCA
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 2721-22-4
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.