Please login to your online account to display your discounted pricing

Alfa Aesar™ Delta Sleep-Inducing Peptide

For cellular and molecular biology applications

Specifications

Product Type Delta Sleep-Inducing Peptide
Molecular Weight 848.81
Storage Requirements -30° to -10°C
Sequence Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu
Format Powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63334MCR

 
alfa aesar™
J63334MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ63334LB0

 
alfa aesar™
J63334LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Delta Sleep-Inducing Peptide
Molecular Weight 848.81
Storage Requirements -30° to -10°C
Sequence Trp-Ala-Gly-Gly-Asp-Ala-Ser-Gly-Glu
Format Powder
Common Name DSIP
Regulatory Status RUO
Purification Method HPLC