Please login to your online account to display your discounted pricing

Alfa Aesar™ delta-Nonanolactone, 98%

$59.22 - $171.77

Chemical Identifiers

Chemical Name or Material delta-Nonanolactone
CAS 3301-94-8
Molecular Formula C9H16O2
Formula Weight 156.23
MDL Number MFCD00036500
Beilstein 114460
Synonym 5-Hydroxynonanoic acid lactone; delta-Nonalactone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1412222

  SDS
alfa aesar™
L1412222
100g Each for $171.77

AAL1412214

  SDS
alfa aesar™
L1412214
25g Each for $59.22
Description & Specifications

Specifications

Chemical Name or Material delta-Nonanolactone
Quantity 100g
CAS 3301-94-8
TSCA TSCA
Assay 98%
Molecular Formula C9H16O2
Formula Weight 156.23
MDL Number MFCD00036500
Beilstein 114460
Synonym 5-Hydroxynonanoic acid lactone; delta-Nonalactone
Boiling Point 250°C
Melting Point −26°C
Flash Point 135°C (275°F)