Please login to your online account to display your discounted pricing

Delavirdine mesylate, Tocris™

Non-nucleoside reverse transcriptase inhibitor

$265.15 - $1,116.26

Chemical Identifiers

Chemical Name or Material Delavirdine mesylate
Assay Percent Range >99%
Molecular Formula C22H28N6O3S·CH3SO3H
Formula Weight 552.67
Synonym 1-[3-[(1-Methylethyl)amino]-2-pyridinyl]-4-[[5-[(methylsulfonyl)amino]-1H-indol-2-yl]carbonyl]-piperazine monomethanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

414950

 
r & d systems
414950
50mg Each for $1,116.26

414910

 
r & d systems
414910
10mg Each for $265.15
Description & Specifications

Specifications

Formula Weight 552.67
Quantity 50mg
Molecular Formula C22H28N6O3S·CH3SO3H
Synonym 1-[3-[(1-Methylethyl)amino]-2-pyridinyl]-4-[[5-[(methylsulfonyl)amino]-1H-indol-2-yl]carbonyl]-piperazine monomethanesulfonate
Chemical Name or Material Delavirdine mesylate
Assay Percent Range >99%
CAS 147221-93-0
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature