Please login to your online account to display your discounted pricing

Dehydroacetic acid sodium salt, 99%, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC364850025

 View more versions of this product

Catalog No. AC364850025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 190.13
Physical Form Powder
Quantity 2.5kg
Color White to yellow
Water 0.5% max. (K.F.)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 4418-26-2
MDL Number MFCD00040583
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Infrared Spectrum Authentic
Packaging Plastic bottle
Synonym 3-Acetyl-6-methyl-2H-pyran-2, 4(3H)-dione sodium salt
Chemical Name or Material Dehydroacetic acid sodium salt
Molecular Formula C8H7O4Na
Name Note 99%

Specifications

Chemical Name or Material Dehydroacetic acid sodium salt
Name Note 99%
CAS 4418-26-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7O4Na
Formula Weight 190.13
MDL Number MFCD00040583
Synonym 3-Acetyl-6-methyl-2H-pyran-2, 4(3H)-dione sodium salt
Accessories

Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents