Learn More
Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 364855000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dehydroacetic acid sodium salt | |
4418-26-2 | |
White to Yellow | |
99% | |
C8H7O4Na | |
500 g | |
190.13 | |
Powder |
99% | |
295.0°C | |
Authentic | |
Plastic bottle | |
MFCD00040583 | |
3-Acetyl-6-methyl-2H-pyran-2, 4(3H)-dione sodium salt | |
99% |
Chemical Identifiers
4418-26-2 | |
C8H7O4Na | |
MFCD00040583 | |
3-Acetyl-6-methyl-2H-pyran-2, 4(3H)-dione sodium salt |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only