missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dehydroacetic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 520-45-6
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00066709
InChI Key PGRHXDWITVMQBC-UHFFFAOYSA-N
Synonym dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
PubChem CID 122903
IUPAC Name 3-acetyl-6-methylpyran-2,4-dione
SMILES CC1=CC(=O)C(C(=O)O1)C(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111940050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
111940050
5g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC111941000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
111941000
100g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC111945000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
111945000
500g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

520-45-6
168.15
PGRHXDWITVMQBC-UHFFFAOYSA-N
122903
CC1=CC(=O)C(C(=O)O1)C(=O)C
C8H8O4
MFCD00066709
dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
3-acetyl-6-methylpyran-2,4-dione

Specifications

520-45-6
Authentic
Glass Bottle
MFCD00066709
17, 559
dehydroacetic acid, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, methylacetopyronone, dehydracetic acid, biocide 470f, acetic acid, dehydro, 3-acetyl-6-methyl-2,4-pyrandione, 2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4
PGRHXDWITVMQBC-UHFFFAOYSA-N
3-acetyl-6-methylpyran-2,4-dione
122903
98%
270.0°C
98%
C8H8O4
5g
15, 2869
Solubility in water: <0.1% (25 c). Other solubilities: 22g/100g acetone- 18g/100g benzene (25 c),5g/100g methanol- 3g/100g ccl4 (25 c),3g/100g ethanol- 0.7g/100g n-heptane (25 c),1.7g/100g propylene glycol- 5g/100g ether (25 c)
CC1=CC(=O)C(C(=O)O1)C(=O)C
168.15
168.15
Dehydroacetic acid, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-293-9

RTECSNumber : UP8050000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only