Learn More
Dehydroacetic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 111940050
Specifications
Dehydroacetic acid | |
Glass Bottle | |
270.0°C | |
109.0°C to 112.0°C | |
98% | |
98% | |
MFCD00066709 | |
15, 2869 | |
Solubility in water: <0.1% (25 c). Other solubilities: 22g/100g acetone- 18g/100g benzene (25 c),5g/100g methanol- 3g/100g ccl4 (25 c),3g/100g ethanol- 0.7g/100g n-heptane (25 c),1.7g/100g propylene glycol- 5g/100g ether (25 c) | |
CC1=CC(=O)C(C(=O)O1)C(=O)C | |
168.15 | |
168.15 | |
98% |
Authentic | |
0.1% max. (700°C, 2 HRS.) | |
Cream or Yellow | |
5g | |
520-45-6 | |
C8H8O4 | |
17, 559 | |
2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-2,4-pyrandione, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4, acetic acid, dehydro, biocide 470f, dehydracetic acid, dehydroacetic acid, methylacetopyronone | |
PGRHXDWITVMQBC-UHFFFAOYSA-N | |
3-acetyl-6-methylpyran-2,4-dione | |
122903 | |
Fine Crystalline Powder |
Chemical Identifiers
520-45-6 | |
168.15 | |
PGRHXDWITVMQBC-UHFFFAOYSA-N | |
122903 | |
CC1=CC(=O)C(C(=O)O1)C(=O)C |
C8H8O4 | |
MFCD00066709 | |
2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl, 3-acetyl-6-methyl-2,4-pyrandione, 3-acetyl-6-methyl-2h-pyran-2,4 3h-dione, 3-acetyl-6-methyl-pyran-2,4-dione, 3-acetyl-6-methyldihydropyrandione-2,4, acetic acid, dehydro, biocide 470f, dehydracetic acid, dehydroacetic acid, methylacetopyronone | |
3-acetyl-6-methylpyran-2,4-dione |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-293-9
RTECSNumber : UP8050000
TSCA : TSCA