Please login to your online account to display your discounted pricing

Alfa Aesar™ Deglycosylated Fetuin Digest Standard, Certified Mass Spec Grade


Manufacturer: alfa aesar™  J67545LPL

 View more versions of this product

Catalog No. AAJ67545LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Format Solid
Quantity 1 vial
For Use With (Application) For the standardizing, calibrating, and troubleshooting chromatographic separations. Particularly useful for LC-MS applications.
Solubility Soluble in water
Storage Requirements -30°C to -10°C
Description White
Product Type Deglycosylated Fetuin