Please login to your online account to display your discounted pricing

Alfa Aesar™ Deglycosylated Fetuin Digest Standard, Certified Mass Spec Grade


Manufacturer: alfa aesar™  J67545ODL

 View more versions of this product

Catalog No. AAJ67545ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Deglycosylated Fetuin
Quantity 3 vials
Storage Requirements -30°C to -10°C
Format Solid
Solubility Soluble in water
Description White
For Use With (Application) For the standardizing, calibrating, and troubleshooting chromatographic separations. Particularly useful for LC-MS applications.