Ohaus™ Defender 5000 Bench Scales, 18 x 24 Inch Platform, Painted Steel Column
Defender 5000 Multifunctional Bench Scale features an indicator that guides the user with on-screen prompts and soft keys guides
Manufacturer:  Ohaus™ 30461639
 View more versions of this product
Catalog No. 01922228
Description
Description
  • Results shown clearly on the large backlit display
  • Alpha-numeric keypad and function keys allow for a simplified user experience
Specifications
Specifications
CE/CSA
18 x 24 in.
Painted Steel Column
AC Adapter
Stainless Steel Pan and Powder Coated Steel Frame Base
2 Seconds
26 kg
250 lb.
125 kg
Standard RS232, MicroSD Card (included). Optional Ethernet, WiFi/Bluetooth, 2nd RS232/RS485/USB, Analog Output, 2 In/4 Out Digital I/O available.
LCD Indicator
23 cm
100 cm
5 g
14°F to 104°F / -10°C to 40°C
68 cm
Legal For Trade
ABS Plastic Indicator
Documents