missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Decyldimethylchlorosilane, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 432190250
Specifications
Decyldimethylchlorosilane | |
0.8660g/mL | |
137°C | |
96% min. (GC) | |
C12H27ClSi | |
25mL | |
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa | |
CCCCCCCCCC[Si](C)(C)Cl | |
234.88 | |
234.88 |
38051-57-9 | |
98°C (3.0 hPa) | |
Authentic | |
Glass bottle | |
1.4400 to 1.4420 (20°C, 589nm) | |
0.866 | |
ZLZGHBNDPINFKG-UHFFFAOYSA-N | |
chloro-decyl-dimethylsilane | |
2758020 | |
97% |
Chemical Identifiers
38051-57-9 | |
234.88 | |
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa | |
chloro-decyl-dimethylsilane |
C12H27ClSi | |
ZLZGHBNDPINFKG-UHFFFAOYSA-N | |
2758020 | |
CCCCCCCCCC[Si](C)(C)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 253-761-8
RUO â Research Use Only