missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Decyldimethylchlorosilane, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 38051-57-9 |
---|---|
Molecular Formula | C12H27ClSi |
Molecular Weight (g/mol) | 234.88 |
InChI Key | ZLZGHBNDPINFKG-UHFFFAOYSA-N |
Synonym | chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa |
PubChem CID | 2758020 |
IUPAC Name | chloro-decyl-dimethylsilane |
SMILES | CCCCCCCCCC[Si](C)(C)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC432190050
|
Thermo Scientific Chemicals
432190050 |
5mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC432190250
|
Thermo Scientific Chemicals
432190250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
38051-57-9 | |
234.88 | |
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa | |
chloro-decyl-dimethylsilane |
C12H27ClSi | |
ZLZGHBNDPINFKG-UHFFFAOYSA-N | |
2758020 | |
CCCCCCCCCC[Si](C)(C)Cl |
Specifications
38051-57-9 | |
98°C (3.0 hPa) | |
Authentic | |
Glass bottle | |
1.4400 to 1.4420 (20°C, 589nm) | |
0.866 | |
ZLZGHBNDPINFKG-UHFFFAOYSA-N | |
chloro-decyl-dimethylsilane | |
2758020 | |
97% |
0.8660g/mL | |
137°C | |
96% min. (GC) | |
C12H27ClSi | |
5mL | |
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa | |
CCCCCCCCCC[Si](C)(C)Cl | |
234.88 | |
234.88 | |
Decyldimethylchlorosilane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 253-761-8
RUO â Research Use Only