Learn More
Decyl sodium sulfate, HPLC grade, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 406220250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ion pair chromatographySpecifications
Decyl sodium sulfate | |
(0.005M in water) (1 cm cell), (0.005 M in water) (1 cm cell) at 0nm, 0.01 max. at 254nm, 0.01 max. at 280nm, 0.05 max. at 220nm, 0.1 max. at 200nm | |
Glass bottle | |
CH3(CH2)9OSO3Na | |
25g | |
Solubility in water: soluble. Other solubilities: soluble in ethanol | |
CCCCCCCCCCOS(=O)(=O)[O-].[Na+] | |
260.32 | |
260.32 | |
HPLC |
142-87-0 | |
Authentic | |
C10H19Na·H2SO4 | |
MFCD00041881 | |
sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt | |
XZTJQQLJJCXOLP-UHFFFAOYSA-M | |
sodium;decyl sulfate | |
23665771 | |
≥98.5% (titrimetric) (on dry substance) |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 205-568-5
RUO â Research Use Only