Please login to your online account to display your discounted pricing

Decyl sodium sulfate, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC406221000

 View more versions of this product

Catalog No. AC406221000

  • / Each
 

Description & Specifications

Chemical Identifiers

Absorbance (0.005 M in water) (1 cm cell)
Chemical Name or Material Decyl sodium sulfate
Quantity 100g
Absorbance Conditions (0.005M in water) (1 cm cell)
CAS 142-87-0
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Appearance of Solution (25% in water at 25°C) clear, colorless to slightly yellow
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Purity Grade HPLC Grade
Physical Form Powder
Assay ≥98.5% (titrimetric) (on dry substance)
Molecular Formula C10H19Na·H2SO4
Linear Formula CH3(CH2)9OSO3Na
Formula Weight 260.32
Infrared Spectrum Authentic
MDL Number MFCD00041881
Synonym Decylsulfuric acid sodium salt
Packaging Glass bottle
Water 1.5% max. (K.F.)
Color White to slightly yellow

Specifications

Chemical Name or Material Decyl sodium sulfate
CAS 142-87-0
Purity Grade HPLC Grade
Assay ≥98.5% (titrimetric) (on dry substance)
Molecular Formula C10H19Na·H2SO4
Linear Formula CH3(CH2)9OSO3Na
Formula Weight 260.32
MDL Number MFCD00041881
Synonym Decylsulfuric acid sodium salt
Accessories

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

SDS & Safety Documents