missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Decyl beta-D-glucopyranoside, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6642303
Description
Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.Specifications
Decyl beta-D-glucopyranoside | |
White | |
MFCD00063297 | |
85089 | |
Soluble in methanol | |
CCCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O | |
320.426 | |
320.42 |
58846-77-8 | |
C16H32O6 | |
1 g | |
decyl beta-d-glucopyranoside, n-decyl-b-d-glucopyranoside, decyl-beta-d-glucopyranoside, n-decyl-beta-d-glucopyranoside, unii-z17h97ea6y, n-decyl glucoside, decyl beta-d-glucoside, triton x 190, 28-deacetylsendanin, beta-d-glucoside, decyl | |
JDRSMPFHFNXQRB-IBEHDNSVSA-N | |
(2R,3R,4S,5S,6R)-2-decoxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
62142 | |
Powder |
Chemical Identifiers
58846-77-8 | |
320.426 | |
JDRSMPFHFNXQRB-IBEHDNSVSA-N | |
62142 | |
CCCCCCCCCCOC1C(C(C(C(O1)CO)O)O)O |
C16H32O6 | |
MFCD00063297 | |
decyl beta-d-glucopyranoside, n-decyl-b-d-glucopyranoside, decyl-beta-d-glucopyranoside, n-decyl-beta-d-glucopyranoside, unii-z17h97ea6y, n-decyl glucoside, decyl beta-d-glucoside, triton x 190, 28-deacetylsendanin, beta-d-glucoside, decyl | |
(2R,3R,4S,5S,6R)-2-decoxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
EINECSNumber : 261-469-7
RTECSNumber : LZ5776531
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only