missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decyl alcohol, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 165760010

 View more versions of this product

Catalog No. AC165760010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Decyl alcohol
6°C
0.8290g/mL
82°C
98% min. (GC)
C10H22O
CH3(CH2)9OH
1 L
0.829
1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
MWKFXSUHUHTGQN-UHFFFAOYSA-N
decan-1-ol
8174
CHEBI:28903
98+%
112-30-1
Colorless to Yellow
220°C to 240°C
Authentic
Glass Bottle
1.4360 to 1.438
MFCD00004747
01, 425
15, 2858
Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,chlorogorm,soluble in alcohol,ether,mineral oil,propylene,insoluble in glycerin water (233°C)
CCCCCCCCCCO
158.28
14.1 mPa.s (20°C)
158.28
Liquid

Chemical Identifiers

112-30-1
158.28
MWKFXSUHUHTGQN-UHFFFAOYSA-N
8174
decan-1-ol
C10H22O
MFCD00004747
1-decanol, decyl alcohol, decanol, n-decyl alcohol, n-decanol, capric alcohol, nonylcarbinol, antak, caprinic alcohol, royaltac
CHEBI:28903
CCCCCCCCCCO
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

EINECSNumber : 203-956-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only