Decon™ SeraSpin Crystals
Plastic pellets used to trap fibrin particles in centrifuged whole blood samples
Manufacturer: Decon Laboratories 2116
Description
- Maximize serum yield in whole blood test samples
- Remove fibrin common in many cardiac patients
- Prevent clotting on instrumentation
- Increase lab efficiency by eliminating need for second centrifuge
Specifications
Crystal | |
Centrifuge | |
Plastic |