Decon™ Proguard™ Professional Hand Cream
Specially formulated lotion moisturizes dry, cracked hands and guards against latex glove irritation
Manufacturer:  Decon™ DC9503
 View more versions of this product
Catalog No. 04-355-20
Description
Description
  • Does not contain grease, oil, perfume, aloe, or other ingredients that can compromise the integrity of latex gloves
  • Absorbs instantly, leaving skin soft
Specifications
Specifications
White
Cream
3 oz.
Tube
Specially formulated lotion moisturizes dry, cracked hands and guards against latex glove irritation
Professional Hand Cream
90mL
Each
Documents