Please login to your online account to display your discounted pricing

Decon™ Neutrad™ Liquid Detergent


For manual scrubbing

Manufacturer: decon laboratories  DC3003

 View more versions of this product

Catalog No. 04-355-10

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Size 32 oz.
Boiling Point 100°C
Certifications/Compliance 29 CFR1910.1200
Color Amber/Straw
Format Liquid
For Use With (Application) For manual scrub cleaning
pH Range 7.2