Please login to your online account to display your discounted pricing
Decon™ ELIMINase™ Decontaminant
Eliminates RNase, DNase, and DNA from laboratory surfaces
Manufacturer: decon laboratories  DC1101
 View more versions of this product
Catalog No. 04-355-31
  • / Each
  • / Case of 12 Each
Description & Specifications
Specifications
Product Type ELIMINase™ Laboratory Decontaminant
For Use With (Application) Eliminates RNase, DNase & DNA contamination
Volume (Metric) 250mL
For Use With (Equipment) Safe on glass, plastics, stainless steel, pipetters, gel boxes, and benchtops
Volume (English) 8.5 oz.