Decon™ Dri-Contrad™ Detergent Powder
Cleans proteins, bloods, oils, waxes, and organic or inorganic contaminants
Manufacturer:  Decon™ DC2002
 View more versions of this product
Catalog No. 04-355-4
Description
Description
  • Nonfoaming powder
  • For automatic washers
  • Free-rinsing, rapidly soluble, and noncorrosive
  • Chlorinated for biohazard control
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Specifications
Specifications
Poly bottle
Solid
4.37 lb.
For automatic washers, removes Proteins, Blood, Oils, Waxes and Organic or Inorganic contaminants
Documents