Please login to your online account to display your discounted pricing

Decon™ Dri-Contrad™ Detergent Powder


Cleans proteins, bloods, oils, waxes, and organic or inorganic contaminants

Manufacturer: decon laboratories  DC2005

 View more versions of this product

Catalog No. 04-355-5

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Specifications

Size 11 lb.
For Use With (Application) For automatic washers, removes Proteins, Blood, Oils, Waxes and Organic or Inorganic contaminants
Format Solid
Container Type Poly bottle