Please login to your online account to display your discounted pricing

Decon™ DeSCAL™ Concentrate


Acidic detergent designed to remove hard-water mineral deposits

Manufacturer: decon laboratories  DC4002

 View more versions of this product

Catalog No. 04-355-45

  • / Each
 

Description & Specifications

Specifications

Product Type Concentrated Descaler
For Use With (Application) Acidic detergent designed to remove hard-water mineral deposits, effectively removes urine scale from animal cages
Volume (English) 5.25 gal.
Volume (Metric) 20L
pH 1
For Use With (Equipment) Use in automatic glassware washers, process equipment, stills, autoclaves and baths