Please login to your online account to display your discounted pricing

Decon™ DeSCAL™ Concentrate


Acidic detergent designed to remove hard-water mineral deposits

Manufacturer: decon laboratories  DC4001*

 View more versions of this product

Catalog No. 04-355-36

  • / Each
  • / Case of 12 Each
 

Description & Specifications

Specifications

Product Type Concentrated Descaler
Volume (English) 34 gal.
For Use With (Equipment) Use in automatic glassware washers, process equipment, stills, autoclaves and baths
Volume (Metric) 1L
pH 1
For Use With (Application) Acidic detergent designed to remove hard-water mineral deposits, effectively removes urine scale from animal cages