Please login to your online account to display your discounted pricing

Decon™ DeSCAL™ Concentrate


Acidic detergent designed to remove hard-water mineral deposits

Manufacturer: decon laboratories  DC4010

 View more versions of this product

Catalog No. 04-355-100

  • / Case of 2 Each
 

Description & Specifications

Specifications

Product Type Concentrated Descaler
For Use With (Application) Acidic detergent designed to remove hard-water mineral deposits, effectively removes urine scale from animal cages
For Use With (Equipment) Use in automatic glassware washers, process equipment, stills, autoclaves and baths
Volume (English) 2.25 gal.
Volume (Metric) 10L
pH 1