Please login to your online account to display your discounted pricing

Decon™ Contrex™ AW Powdered Autowasher Detergent


Formulated for use in all automatic labware washing machines

Manufacturer: decon laboratories  DC5604

 View more versions of this product

Catalog No. 04-358-12

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Specifications

Format Powder
Size 4 lb.
Color White
Container Type Poly bottle
For Use With (Application) For use in all automatic labware washing machines
pH Range 11.5 to 12.5