Please login to your online account to display your discounted pricing

Decon™ Contrex™ AW Powdered Autowasher Detergent


Formulated for use in all automatic labware washing machines

Manufacturer: decon laboratories  DC5650

 View more versions of this product

Catalog No. 04-358-14

  • / Each
 

Description & Specifications

Specifications

Size 50 lb.
Color White
Format Powder
For Use With (Application) For use in all automatic labware washing machines
pH Range 11.5 to 12.5