Decon™ Contrex™ AP Powdered Labware Detergent
Ideal for cleaning labware, scientific apparatus, and process equipment
Manufacturer:  Decon Laboratories DC5225
 View more versions of this product
Catalog No. 04-358-18
Specifications Description & Specifications
Specifications
25 lb.
For use in manual, soak or ultrasonic cleaning; Cleans labware, scientific apparatus and process equipment
White
Powder
9.5 to 11