Decon™ Contrex™ AL Alkaline Liquid Detergent
For use in manual, soak, or ultrasonic cleaning
Supplier: Decon™ 5301
Description
For use in manual, soak, or ultrasonic cleaning
Ideal for cleaning labware, scientific apparatus, and process equipment.
- Concentrated liquid
- Free-rinsing, phosphate-free, and disposable straight to drain
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
- Non-toxic: Item is non-toxic
Specifications
7.5 to 9.5 | |
Pale Yellow | |
For use in manual, soak or ultrasonic cleaning of labware, scientific apparatus and process equipment | |
3.8L |
29 CFR1910.1200 | |
Liquid | |
1 gal. |