Please login to your online account to display your discounted pricing

Decon™ Contrad™ NF Liquid Detergent


Cleans a wide variety of organic and inorganic contaminants from most substrates

Manufacturer: decon laboratories  DC6001*

 View more versions of this product

Catalog No. 04-355-30

  • / Case of 2 Each
 

Description & Specifications

Specifications

Size 2.62 gal.
Boiling Point >100°C
Format Liquid
For Use With (Application) For automatic washing machines, removes Organic and Inorganic contaminants
pH Range 14