Decon™ Contrad™ NF Liquid Detergent
Cleans a wide variety of organic and inorganic contaminants from most substrates
Manufacturer:  Decon™ DC6055
 View more versions of this product
Catalog No. 04-355-40
Description
Description
  • Super-concentrated alkaline liquid
  • Formulated specifically for automatic washing machines
  • Nonfoaming, free-rinsing, and residue-free
  • Phosphate- and chlorine-free
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic
Specifications
Specifications
>100°C
55.5 gal.
14
Liquid
For automatic washing machines, removes Organic and Inorganic contaminants
Documents