Please login to your online account to display your discounted pricing

Decon™ Contrad™ 70 Liquid Detergent


Cleans glass, plastic, ceramic, and ferrous metals by soaking, scrubbing, or ultrasonic cleaning

Manufacturer: decon laboratories  DC1005

 View more versions of this product

Catalog No. 04-355-3

  • / Each
 

Description & Specifications

Specifications

Boiling Point 100°C
Certifications/Compliance USDA, 29 CFR 1910.1200
Size 55.5 gal.
pH Range 13
Color White
Format Liquid