Please login to your online account to display your discounted pricing

Decon™ Contrad™ 100 Equipment Cleaner


Formulated for clean-in-place systems and high-pressure cleaning applications

Manufacturer: decon laboratories  DC1505

 View more versions of this product

Catalog No. 04-355-57

  • / Each
 

Description & Specifications

Specifications

Format Liquid
Boiling Point 100°C
pH Range 12.4
Sterile Yes
Size 55.5 gal.
For Use With (Application) For clean in place systems and high pressure cleaning applications
Color Clear