Please login to your online account to display your discounted pricing

Decon™ Contrad™ 100 Equipment Cleaner


Formulated for clean-in-place systems and high-pressure cleaning applications

Manufacturer: decon laboratories  DC1503

 View more versions of this product

Catalog No. 04-355-55

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) For clean in place systems and high pressure cleaning applications
Format Liquid
Sterile Yes
pH Range 12.4
Size 5.25 gal.
Boiling Point 100°C
Color Clear