missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decon™ Conflikt™ Detergent Disinfectant

Prediluted ready-to-use spray quaternary ammonium disinfectant

Manufacturer:  Decon™ DC4102

 View more versions of this product

Catalog No. 04-355-34




Description

Description

Effective against TB, HBV, and a wide range of viruses, bacteria, pathogenic fungi, mold, and mildew, including HIV-1, E. coli, and Canine Parvovirus.

  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic

Certifications

USDA approved

Compliance

  • EPA registered
  • Meets OSHA's Bloodborne Pathogen Standard

Specifications

Specifications

200°F
Clear to Light Yellow
Trigger spray bottle
Liquid
32 oz.
11.7
EPA, OSHA, USDA
0.105% n-Alkyl Dimethyl Benzyl Ammonium Chlorides, 0.105% n-Alkyl Dimethyl Ethylbenzyl Ammonium Chlorides, 99.790% inert ingredients
Cleans and disinfects hard surfaces that may harbor Viruses, Bacteria and Fungi
50° to 120°F
946mL
Documents