Decon™ Conflikt™ Detergent Disinfectant
Prediluted ready-to-use spray quaternary ammonium disinfectant
Manufacturer:  Decon Laboratories DC4103
 View more versions of this product
Catalog No. 04-355-35
Specifications Description & Specifications
Specifications
Clear to Light Yellow
Cleans and disinfects hard surfaces that may harbor Viruses, Bacteria and Fungi
<50 or >120°F
11.7
200°F
EPA, OSHA, USDA
Bottle
64 oz.
0.105% n-Alkyl Dimethyl Benzyl Ammonium Chlorides, 0.105% n-Alkyl Dimethyl Ethylbenzyl Ammonium Chlorides, 99.790% inert ingredients
1.9L
64 oz.
Liquid