missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decon™ CiDehol™ ST Sterile 70% Isopropyl Alcohol Solution made with WFI

Sterile, ready-to-use 70% isopropyl alcohol (IPA) solution designed especially for cleanroom use. Decon Labs, CiDehol™ ST Sterile 70% Isopropyl Alcohol Solution made with WFI is prepared according to USP specifications and packaged in Class 100 cleanroom.

Manufacturer:  Decon Laboratories DC8301

 View more versions of this product

Catalog No. 04-355-80


 

Specifications

Description

Spray; CiDehol ST; 1gal. (3.8L); 70% isopropyl alcohol (IPA) spray; Filtered to 0.2 micron; Double-bagged in an ISO Class 5 environment; Sterilized via gamma irradiation

  • Made with Water-For-Injection
  • Solution is filtered to 0.2μm and individually double-bagged in an ISO Class 5 (Class 100) environment, then sterilized via gamma irradiation
  • Each lot undergoes sterility and pyrogen testing and meets WFI limits in endotoxin
  • Each case shipped with a lot-specific document, which details QC, irradiation, and testing

Description & Specifications

Specifications

Yes
USP, ISO 5, EPA
CiDehol™ ST Spray
70% Isopropyl Alcohol, 30% Water for Injection
Liquid
70% Isopropyl Alcohol
Designed especially for cleanroom use
1 gal.
0.2μm
Hands-free dispenser
1 gal., 3.8L
3.8L