Decon™ AquaPur™ Purified USP-Grade Water
For use in laboratory or production applications
Supplier:  Decon™ DC9301
 View more versions of this product
Catalog No. 04-355-124
Alert:
Not to be used for parenteral administration. Not for use in sterile irrigation, injection, infusion, or as a potable product.For manufacturing and laboratory use only.
Description
Description
Tested to meet USP specifications and packaged in convenient point-of-use packaging. Proprietary manufacturing process ensures AquaPur Purified Water maintains microbial specification through expiration date.
Meets these specifications:
  • USP Water Conductivity, USP 645 (at manufacture)
  • USP Total Organic Carbon, USP 643 < 0.5ppm
  • Oxidizable Substances: Conforms (at manufacture)
  • Total Microbial Count: < 100cfu/mL
Specifications
Specifications
1 gal.