Please login to your online account to display your discounted pricing

Decon™ AquaPur™ Purified USP-Grade Water


For use in laboratory or production applications

Manufacturer: decon laboratories  DC9305

 View more versions of this product

 Alert

Not to be used for parenteral administration. Not for use in sterile irrigation, injection, infusion, or as a potable product.For manufacturing and laboratory use only.

Catalog No. 04-355-125

  • / Each
 

Description & Specifications

Specifications

Boiling Point 100°C
Format Liquid
For Use With (Application) For use in laboratory or production applications requiring purified water
Size 5 gal.
Certifications/Compliance CAP/NCCLS Type I, USP
Color Colorless