Please login to your online account to display your discounted pricing

Alfa Aesar™ Decanophenone, 98+%

$57.57 - $159.72

Chemical Identifiers

Chemical Name or Material Decanophenone
CAS 6048-82-4
Molecular Formula C16H24O
Formula Weight 232.37
MDL Number MFCD00009582
Beilstein 2048790
Synonym n-Nonyl phenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0692306

  SDS
alfa aesar™
L0692306
5g Each for $57.57

AAL0692314

  SDS
alfa aesar™
L0692314
25g Each for $159.72
Description & Specifications

Specifications

Formula Weight 232.37
Chemical Name or Material Decanophenone
MDL Number MFCD00009582
CAS 6048-82-4
TSCA TSCA
Assay ≥98%
Molecular Formula C16H24O
Boiling Point 165° to 167°C (7mmHg)
Melting Point 34° to 36°C
Beilstein 2048790
Quantity 5g
Flash Point >110°C (230°F)
Synonym n-Nonyl phenyl ketone